Anti-RGS21 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-11620R-A555
10263-436EA 811.1 USD
10263-436
Anti-RGS21 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
The regulators of G protein signaling (RGS) proteins inhibit heterotrimeric G protein signaling. RGS proteins work by functioning as GTPase-activators (which increase the GTPase activity of G protein a-subunits) thereby driving G proteins into their inactive GDP-bound form. RGS21 (regulator of G-protein signaling 21) is a 152 amino acid protein that is ubiquitously expressed and contains one RGS domain. The gene encoding RGS21 maps to human chromosome 1q31.2 and mouse chromosome 1 F. Human chromosome 1 spans 260 million base pairs, contains over 3,000 genes, comprises nearly 8% of the human genome and houses a large number of disease-associated genes, including those that are involved in familial adenomatous polyposis, Stickler syndrome, Parkinson’s disease, Gaucher disease, schizophrenia and Usher syndrome.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RGS21
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR