Anti-GRM2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-1161R-A555
10260-140EA 811.1 USD
10260-140
Anti-GRM2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
G-protein coupled receptor for glutamate. Ligand binding causes a conformation change that triggers signaling via guanine nucleotide-binding proteins (G proteins) and modulates the activity of down-stream effectors, such as adenylate cyclase. Signaling inhibits adenylate cyclase activity. May mediate suppression of neurotransmission or may be involved in synaptogenesis or synaptic stabilization.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Metabotropic glutamate receptor 2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR