Anti-NXPH1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-11170R-HRP
10250-506EA 811.1 USD
10250-506
Anti-NXPH1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
May be signaling molecules that resemble neuropeptides and that act by binding to alpha-neurexins and possibly other receptors.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: NXPH1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR