Anti-RARRES2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-10410R-A350
10243-438EA 811.1 USD
10243-438
Anti-RARRES2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Chemerin, also known as retinoic acid receptor responder 2 (RARRES2) and tazarotene induced gene 2 (TIG2), is a chemoattractant protein for cells expressing G-protein-linked receptor chemokine-like receptor 1 (CMKLR1, ChemR23 or GPCR-DEZ). The protein is synthesized as a secreted precursor, prochemerin (18 kDa), which is poorly active. Upon proteolytic removal of the last six amino acids, prochemerin is converted into mature active chemerin (15-16 kDa) that acts as a strong and highly specific agonist of its receptor CMKLR1. Chemerin is a newly described adipokine with effects on adipocyte differnetiation and metabolism.

Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: RARRES2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR