Anti-F5 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-1040R-A488
10243-416EA 810.85 USD
10243-416
Anti-F5 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Central regulator of hemostasis. It serves as a critical cofactor for the prothrombinase activity of factor Xa that results in the activation of prothrombin to thrombin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: F5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR