Anti-CD4 Rabbit Polyclonal Antibody

Supplier: Bioss

BS-0647R
10238-584EA 651.79 USD
10238-584
Anti-CD4 Rabbit Polyclonal Antibody
Antibodies
Accessory protein for MHC class-II antigen/T-cell receptor interaction. May regulate T-cell activation.

Recommended Dilutions: Western Blot: 1:100-1000; Flow Cytometry: 1:20-100; IHC-P: 1:100-500; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD4
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR