Anti-CD34 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-0646R-CY3
10238-572EA 811.1 USD
10238-572
Anti-CD34 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Possible adhesion molecule with a role in early hematopoiesis by mediating the attachment of stem cells to the bone marrow extracellular matrix or directly to stromal cells. Could act as a scaffold for the attachment of lineage specific glycans, allowing stem cells to bind to lectins expressed by stromal cells or other marrow components. Presents carbohydrate ligands to selectins.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CD34
Clonality: Polyclonal
Clone: 0
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR