Anti-Thioster-containing protein 1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-0632R-CY5.5
10239-000EA 811.1 USD
10239-000
Anti-Thioster-containing protein 1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
No Data Available.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Thioster-containing protein 1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR