Anti-Thioster-containing protein 1 Rabbit Polyclonal Antibody (Biotin)

BS-0632R-BIO
10238-994EA 529.29 USD
10238-994
Anti-Thioster-containing protein 1 Rabbit Polyclonal Antibody (Biotin)
No Data Available.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: Thioster-containing protein 1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR