Anti-Thioster-containing protein 1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-0632R-A350
10238-986EA 811.1 USD
10238-986
Anti-Thioster-containing protein 1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
No Data Available.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Thioster-containing protein 1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Other Species
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR