ZNF300 contains C2H2 (or Kruppel)-type zinc finger motifs that tetrahedrally coordinate zinc ions and form a protein structure that can bind DNA and regulate transcription (Gou et al.,2004 [PubMed 14746915]). ZNF300 belongs to the krueppel C2H2 type zinc finger protein family. It contains twelve C2H2 type zinc fingers and one KRAB domain. ZNF300 has a transcriptional repressor activity. Expressed mostly in heart, skeletal muscle and brain. Isoforms 1 and 2 are highly expressed in testis. There are three named isoforms.
Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500
Type: Primary
Antigen: ZNF300
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human