Anti-ZNF300 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-0628R-A555
10238-918EA 746.42 USD
10238-918
Anti-ZNF300 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
ZNF300 contains C2H2 (or Kruppel)-type zinc finger motifs that tetrahedrally coordinate zinc ions and form a protein structure that can bind DNA and regulate transcription (Gou et al.,2004 [PubMed 14746915]). ZNF300 belongs to the krueppel C2H2 type zinc finger protein family. It contains twelve C2H2 type zinc fingers and one KRAB domain. ZNF300 has a transcriptional repressor activity. Expressed mostly in heart, skeletal muscle and brain. Isoforms 1 and 2 are highly expressed in testis. There are three named isoforms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ZNF300
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR