Anti-CMTM2 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-0265R-CY5
10234-202EA 776.17 USD
10234-202
Anti-CMTM2 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
CKLFSF2 isoform 1

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CMTM2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR