Anti-EPHB2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-0247R-A350
10233-944EA 776.17 USD
10233-944
Anti-EPHB2 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Eph receptors play important roles in axon guidance at the midline. In the auditory system, growth of axons across the midline is an important determinant of auditory function. The avian cochlear nucleus, n. magnocellularis (NM), makes bilateral projections to its target, n. laminaris (NL).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EPHB2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR