Anti-SHANK1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-0211R-CY5
10233-302EA 776.17 USD
10233-302
Anti-SHANK1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Seems to be an adapter protein in the postsynaptic density (PSD) of excitatory synapses that interconnects receptors of the postsynaptic membrane including NMDA-type and metabotropic glutamate receptors via complexes with GKAP/PSD-95 and Homer, respectively, and the actin-based cytoskeleton. Plays a role in the structural and functional organization of the dendritic spine and synaptic junction.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SHANK1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR