Anti-RTN4 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-0134R-CY5
10232-128EA 776.17 USD
10232-128
Anti-RTN4 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Developmental neurite growth regulatory factor with a role as a negative regulator of axon-axon adhesion and growth, and as a facilitator of neurite branching. Regulates neurite fasciculation, branching and extension in the developing nervous system. Involved in down-regulation of growth, stabilization of wiring and restriction of plasticity in the adult CNS. Regulates the radial migration of cortical neurons via an RTN4R-LINGO1 containing receptor complex. May inhibit BACE1 activity and amyloid precursor protein processing.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RTN4
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR