Anti-AR Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-0118R-CY5
10231-748EA 776.17 USD
10231-748
Anti-AR Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Steroid hormone receptors are ligand-activated transcription factors that regulate eukaryotic gene expression and affect cellular proliferation and differentiation in target tissues. Transcription factor activity is modulated by bound coactivator and corepressor proteins. Transcription activation is down-regulated by NR0B2. Activated, but not phosphorylated, by HIPK3 and ZIPK/DAPK3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AR
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR