Anti-SPP1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-0026R-CY3
10229-552EA 811.1 USD
10229-552
Anti-SPP1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Binds tightly to hydroxyapatite. Appears to form an integral part of the mineralized matrix. Probably important to cell-matrix interaction. Acts as a cytokine involved in enhancing production of interferon-gamma and interleukin-12 and reducing production of interleukin-1 and is essential in the pathway that leads to type I immunity.

Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200

Type: Primary
Antigen: SPP1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR