Anti-GSK3B Mouse Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss

BS-0023M-FITC
10229-080EA 811.1 USD
10229-080
Anti-GSK3B Mouse Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
The protein encoded by this gene is a serine-threonine kinase, belonging to the glycogen synthase kinase subfamily. It is involved in energy metabolism, neuronal cell development, and body pattern formation. Polymorphisms in this gene have been implicated in modifying risk of Parkinson disease, and studies in mice show that overexpression of this gene may be relevant to the pathogenesis of Alzheimer disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, Sep 2009]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GSK-3 beta NT
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR