Anti-OPN Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-0019R-A488
10229-384EA 811.1 USD
10229-384
Anti-OPN Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Acts as a cytokine involved in enhancing production of interferon-gamma and interleukin-12 and reducing production of interleukin-10 and is essential in the pathway that leads to type I immunity (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: OPN
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR