Anti-IGF2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-0015R-A555
10229-266EA 811.1 USD
10229-266
Anti-IGF2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
The insulin-like growth factors possess growth-promoting activity. In vitro, they are potent mitogens for cultured cells. IGF-II is influenced by placental lactogen and may play a role in fetal development. Preptin undergoes glucose-mediated co-secretion with insulin, and acts as physiological amplifier of glucose-mediated insulin secretion. Exhibits osteogenic properties by increasing osteoblast mitogenic activity through phosphoactivation of MAPK1 and MAPK3.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IGF2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR