|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | IGF1 |
Antigen Name | Insulin Like Growth Factor 1 |
Reactivity | Mouse, Rat, Human |
Conjugation | Cy5® |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
Antigen Synonyms | IGF-I, MGF, Mechano growth factor, Somatomedin-C, IGFI, Insulin-like growth factor I |
Gene ID | 3479 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Immunogen | 67-90/195 |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |