Anti-IGF1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-0014R-A488
10229-224EA 811.1 USD
10229-224
Anti-IGF1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
The insulin-like growth factors, isolated from plasma, are structurally and functionally related to insulin but have a much higher growth-promoting activity. May be a physiological regulator of [1-14C]-2-deoxy-D-glucose (2DG) transport and glycogen synthesis in osteoblasts. Stimulates glucose transport in rat bone-derived osteoblastic (PyMS) cells and is effective at much lower concentrations than insulin, not only regarding glycogen and DNA synthesis but also with regard to enhancing glucose uptake. May play a role in synapse maturation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: IGF1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR