Anti-SNCA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-0009R-HRP
10229-042EA 811.1 USD
10229-042
Anti-SNCA Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
May be involved in the regulation of dopamine release and transport. Induces fibrillization of microtubule-associated protein tau. Reduces neuronal responsiveness to various apoptotic stimuli, leading to a decreased caspase-3 activation.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: SNCA
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR