Anti-SNCA Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-0009R-CY3
10229-152EA 811.1 USD
10229-152
Anti-SNCA Rabbit Polyclonal Antibody (Cy3®)
Antibodies
May be involved in the regulation of dopamine release and transport. Induces fibrillization of microtubule-associated protein tau. Reduces neuronal responsiveness to various apoptotic stimuli, leading to a decreased caspase-3 activation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SNCA
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR