|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | SNCA |
Antigen Name | Synuclein, Alpha |
Reactivity | Mouse, Rat, Human |
Conjugation | Biotin |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
ImmunoChemistry | Yes |
Antigen Synonyms | PARK1, Non-A beta component of AD amyloid, PD1, Non-A4 component of amyloid precursor, PARK4, NACP |
Gene ID | 6622 |
Concentration | 1 μg/μl |
Cross Adsorption | No |
Immunogen | 45-95/140 |
Purification | Purified by Protein A |
Form | liquid |
Storage Buffer | Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months. |
Storage Temperature | Store at 4°C for 12 months |
Shipping Temperature | 4°C |