Anti-ADM Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-0007R-A350
10228-996EA 811.1 USD
10228-996
Anti-ADM Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
AM and PAMP are potent hypotensive and vasodilatator agents. Numerous actions have been reported most related to the physiologic control of fluid and electrolyte homeostasis. In the kidney, am is diuretic and natriuretic, and both am and pamp inhibit aldosterone secretion by direct adrenal actions. In pituitary gland, both peptides at physiologically relevant doses inhibit basal ACTH secretion. Both peptides appear to act in brain and pituitary gland to facilitate the loss of plasma volume, actions which complement their hypotensive effects in blood vessels.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR