Anti-TSHR Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-0003R-A488
10228-972EA 746.42 USD
10228-972
Anti-TSHR Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Receptor for thyrothropin. Plays a central role in controlling thyroid cell metabolism. The activity of this receptor is mediated by G proteins which activate adenylate cyclase. Also acts as a receptor for thyrostimulin (GPA2+GPB5).

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: TSHR
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR