Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HandiMark® Printer Labels, Brady®

Supplier: Brady Worldwide
142297 142296 142299 142298 142271 142270 142273 142272 142301 142300 142303 142302 142269 142304 42068 42101 42069 42102 42103 42104 42064 42065 42066 42100 42067 142280 142282 42105 142281 142284 142283 142275 142274 142277 142276 142279 142278 42062 42063 42013 42014 42015 42011 42012 142291 142290 142293 142292 142295 142294 142286 142285 142288 142287 142289
General Labels for Straws, Petri Dishes, Bottles, Flasks, Microplates, Bags, and more.



 

ORDER

Expand All / Collapse All

B-595 General Purpose Label for Drawers, Shelves, and Floor
Size Material For Use With Color Supplier No. VWR Catalog Number Unit Price Quantity
0.500" x 50' Vinyl HandiMark®, BMP®71 Black 142287 89205-042 Each Retrieving
1" x 50' Vinyl HandiMark®, BMP®71 Black 142288 89205-044 Each Retrieving
2" x 50' Vinyl HandiMark®, BMP®71 Black 142289 89205-046 Each Retrieving
0.500" x 50' Vinyl HandiMark®, BMP®71 Blue 142281 89205-048 Each Retrieving
1" x 50' Vinyl HandiMark®, BMP®71 Blue 142282 89205-050 Each Retrieving
2" x 50' Vinyl HandiMark®, BMP®71 Blue 142283 89205-052 Each Retrieving
0.500" x 50' Vinyl HandiMark®, BMP®71 Brown 142293 89205-054 Each Retrieving
1" x 50' Vinyl HandiMark®, BMP®71 Brown 142294 89205-056 Each Retrieving
2" x 50' Vinyl HandiMark®, BMP®71 Brown 142295 89205-058 Each Retrieving
0.500" x 50' Vinyl HandiMark®, BMP®71 Clear 142302 89205-060 Each Retrieving
1" x 50' Vinyl HandiMark®, BMP®71 Clear 142303 89205-062 Each Retrieving
2" x 50' Vinyl HandiMark®, BMP®71 Clear 142304 89205-064 Each Retrieving
0.500" x 50' Vinyl HandiMark®, BMP®71 Gold 142296 89205-066 Each Retrieving
1" x 50' Vinyl HandiMark®, BMP®71 Gold 142297 89205-068 Each Retrieving
2" x 50' Vinyl HandiMark®, BMP®71 Gold 142298 89205-070 Each Retrieving
0.500" x 50' Vinyl HandiMark®, BMP®71 Green 142275 89205-072 Each Retrieving
1" x 50' Vinyl HandiMark®, BMP®71 Green 142276 89205-074 Each Retrieving
2" x 50' Vinyl HandiMark®, BMP®71 Green 142277 89205-076 Each Retrieving
0.500" x 50' Vinyl HandiMark®, BMP®71 Grey 142290 89205-078 Each Retrieving
1" x 50' Vinyl HandiMark®, BMP®71 Grey 142291 89205-080 Each Retrieving
2" x 50' Vinyl HandiMark®, BMP®71 Grey 142292 89205-082 Each Retrieving
0.500" x 50' Vinyl HandiMark®, BMP®71 Orange 142284 89205-086 Each Retrieving
1" x 50' Vinyl HandiMark®, BMP®71 Orange 142285 89205-088 Each Retrieving
2" x 50' Vinyl HandiMark®, BMP®71 Orange 142286 89205-090 Each Retrieving
0.500" x 50' Vinyl HandiMark®, BMP®71 Purple 142299 89205-092 Each Retrieving
1" x 50' Vinyl HandiMark®, BMP®71 Purple 142300 89205-094 Each Retrieving
2" x 50' Vinyl HandiMark®, BMP®71 Purple 142301 89205-096 Each Retrieving
0.500" x 50' Vinyl HandiMark®, BMP®71 Red 142278 89205-098 Each Retrieving
1" x 50' Vinyl HandiMark®, BMP®71 Red 142279 89205-100 Each Retrieving
2" x 50' Vinyl HandiMark®, BMP®71 Red 142280 89205-102 Each Retrieving
0.500" x 50' Vinyl HandiMark® White 142269 89205-104 Each Retrieving
1" x 50' Vinyl HandiMark® White 142270 89205-106 Each Retrieving
2" x 50' Vinyl HandiMark® White 142271 89205-108 Each Retrieving
0.500" x 50' Vinyl HandiMark®, BMP®71 Yellow 142272 89205-110 Each Retrieving
1" x 50' Vinyl HandiMark®, BMP®71 Yellow 142273 89205-112 Each Retrieving
2" x 50' Vinyl HandiMark®, BMP®71 Yellow 142274 89205-114 Each Retrieving

B-529 Polypropylene Film with Acrylic Pressure Sensitive Adhesive
Size Material For Use With Color Supplier No. VWR Catalog Number Unit Price Quantity
0.500" x 50' Polypropylene HandiMark®, BMP®71 White 42068 12222-146 Each Retrieving
1" x 50' Polypropylene HandiMark®, BMP®71 White 42069 12222-570 Each Retrieving

B-581 Indoor/Outdoor High Gloss Flexible Pressure Sensitive
Size Material For Use With Color Supplier No. VWR Catalog Number Unit Price Quantity
0.500" x 50' Vinyl HandiMark®, BMP®71 White 42100 12222-540 Each Retrieving
1" x 50' Vinyl HandiMark®, BMP®71 White 42101 12222-574 Each Retrieving
2" x 50' Vinyl HandiMark®, BMP®71 White 42102 12222-532 Each Retrieving
0.500" x 50' Vinyl HandiMark®, BMP®71 Yellow 42103 12222-538 Each Retrieving
1" x 50' Vinyl HandiMark®, BMP®71 Yellow 42104 12222-614 Each Retrieving
2" x 50' Vinyl HandiMark®, BMP®71 Yellow 42105 12222-534 Each Retrieving

B-584 Reflective Tape
Size Material For Use With Color Supplier No. VWR Catalog Number Unit Price Quantity
1" x 30' Retroreflective Tape HandiMark®, BMP®71 Reflective Orange 42066 12222-622 Each Retrieving
2" x 30' Retroreflective Tape HandiMark®, BMP®71 Reflective Orange 42067 12222-628 Each Retrieving
1" x 30' Retroreflective Tape HandiMark®, BMP®71 Reflective Silver 42062 12222-618 Each Retrieving
2" x 30' Retroreflective Tape HandiMark®, BMP®71 Reflective Silver 42063 12222-624 Each Retrieving
1" x 33' Retroreflective Tape HandiMark®, BMP®71 Reflective Yellow 42064 12222-620 Each Retrieving
2" x 33' Retroreflective Tape HandiMark®, BMP®71 Reflective Yellow 42065 12222-626 Each Retrieving

Other HandiMark® Printer Labels
Size For Use With Color Supplier No. VWR Catalog Number Unit Price Quantity
2" x 75' HandiMark® Black 42011 12222-672 Each Retrieving
2" x 75' HandiMark® Blue 42014 12222-666 Each Retrieving
2" x 75' HandiMark® Green 42015 12222-664 Each Retrieving
2" x 75' HandiMark® Red 42013 12222-670 Each Retrieving
2" x 75' HandiMark® White 42012 12222-668 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping