Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HyFlex® 11-539 Ultralight Weight, 18-Gauge Gloves, Fully Coated, Ansell

Supplier: Ansell Healthcare
HyFlex®
11539110 11539100 11539080 11539090 11539060 11539070
Offering exceptional grip performance and fully coated protection in a comfortable, ultralight weight, cut resistant glove, the HyFlex® 11-539 epitomizes the convergence of protection and performance.

  • Enhanced abrasion resistance
  • Excellent grip
  • Superior tactile sensitivity
  • Unparalleled dexterity

It features an 18G ultra-lightweight liner with INTERCEPT TECHNOLOGY® yarns that provides EN Cut Level 3 (ANSI Cut Level 2) protection

This glove's ultralight weight enables unparalleled dexterity and refined mobility for bare hand-like tactility. ZONZ™ comfort fit technology uses selected yarns and varying stitch designs to tailor overall glove fit to enhance hand movement for higher dexterity and reduced fatigue. Its silicone-free design is paint- and finish-process-friendly and dirt-masking palm allows longer wear times.

INTERCEPT TECHNOLOGY® yarn has been designed to perform at or to exceed industry standards. This Ansell proprietary technology offers best-in-class cut protection and increased comfort with dexterity. The 11-539 glove features a fully coated back-of-hand for comprehensive protection against snags and mechanical aggression. ANSELL GRIP™ technology is a coating treatment that minimizes the force required to grip oily or wet tools or materials, reducing hand and arm fatigue and improving control, safety and productivity. The HyFlex® 11-539 glove also features FORTIX™ technology which applies a thin, resilient and breathable nitrile foam coating, which extends gloves’ working life and improves comfort in abrasive applications.

For use in the aerospace, automotive, machinery and equipment, and metal fabrication industries for assembly of small sharp parts, handling small sharp metal parts, light duty stamping and maintenance.

Certifications: ANSI Cut Level 2. ANSI Abrasion Level 4.



 

ORDER

Expand All / Collapse All
Glove Size Length Glove Color Supplier No. VWR Catalog Number Unit Price Quantity
6 22 to 27 cm (8 11/16 to 10 5/8") Gray 11539060 10001-996 Case of 12 (12Pair) Retrieving
Pack of 12 Retrieving
7 22 to 27 cm (8 11/16 to 10 5/8") Gray 11539070 10002-058 Case of 12 (12Pair) Retrieving
Pack of 12 Retrieving
8 22 to 27 cm (8 11/16 to 10 5/8") Gray 11539080 10002-060 Case of 12 (12Pair) Retrieving
Pack of 12 Retrieving
9 22 to 27 cm (8 11/16 to 10 5/8") Gray 11539090 10002-062 Case of 12 (12Pair) Retrieving
Pack of 12 Retrieving
10 22 to 27 cm (8 11/16 to 10 5/8") Gray 11539100 10002-064 Case of 12 (12Pair) Retrieving
Pack of 12 Retrieving
11 22 to 27 cm (8 11/16 to 10 5/8") Gray 11539110 10002-066 Case of 12 (12Pair) Retrieving
Pack of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Ambidextrous/Hand-specific Hand-Specific
Disposable/Reusable Reusable
Liner Material INTERCEPT TECHNOLOGY® yarn and Synthetic Blend
Finish Fully dipped
Cuff Style Knit Wrist
EN Norm 4342

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty