Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HyFlex® 11-727 Medium Duty, 15-Gauge Gloves, Palm Dipped, Ansell

Supplier: Ansell Healthcare
HyFlex®
163837 163838 163836 163840 163841 163839
The HyFlex® 11-727 glove features a 15G lightweight, medium-duty liner with INTERCEPT TECHNOLOGY® yarns that provide EN Cut Level 3 (ANSI Level 2) protection.

  • Silicone-free
  • Increased worker confidence when handling small, sharp parts
  • Exceptional abrasion resistance
  • Minimal shrinkage after laundering

The thin, ergonomic liner features an optimized Lycra® that is soft and cool on the skin for all-day comfort

Precision, dexterity, and flexibility when handling small, sharp parts increases worker confidence while on the job. Durable and launderable with minimal shrinkage and excellent recovery after laundering, extending glove life.

INTERCEPT TECHNOLOGY® yarn has been tested to perform at or exceed industry standards. This Ansell proprietary technology offers best-in-class cut protection and increased comfort and dexterity. The palm coating design improves worker grip, extends glove life in abrasive conditions and allows the back of hand to breathe.

For use in the aerospace, automotive, machinery and equipment, and metal fabrication industries for assembly of small sharp parts, handling small sharp metal parts, light-duty stamping and maintenance.

Certifications: ANSI Cut Level 2. ANSI Abrasion Level 6.



 

ORDER

Expand All / Collapse All
Glove Size Length Glove Color Supplier No. VWR Catalog Number Unit Price Quantity
6 20 to 25.5 cm (7 7/8 to 10") Gray 163836 10001-968 Case of 12 (12Pair) Retrieving
Pack of 12 Retrieving
7 20 to 25.5 cm (7 7/8 to 10") Gray 163837 10001-970 Case of 12 (12Pair) Retrieving
Pack of 12 Retrieving
8 20 to 25.5 cm (7 7/8 to 10") Gray 163838 10001-972 Case of 12 (12Pair) Retrieving
Pack of 12 Retrieving
9 20 to 25.5 cm (7 7/8 to 10") Gray 163839 10001-974 Case of 12 (12Pair) Retrieving
Pack of 12 Retrieving
10 20 to 25.5 cm (7 7/8 to 10") Gray 163840 10001-976 Case of 12 (12Pair) Retrieving
Pack of 12 Retrieving
5 22 to 27 cm (8 11/16 to 10 5/8") Gray 163841 10002-094 Case of 12 (12Pair) Retrieving
Pack of 12 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Ambidextrous/Hand-specific Hand-Specific
Disposable/Reusable Reusable
Liner Material INTERCEPT TECHNOLOGY® yarn and Lycra®/Nylon INTERCEPT TECHNOLOGY® yarn and Synthetic Blend INTERCEPT TECHNOLOGY® yarn and Lycra®/Nylon
Finish Palm Coated Palm dipped Palm Coated
Cuff Style Knit Wrist
EN Norm 4342 4421 4342

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty