The ImmunoReagents' DyLight 680 produces near-infrared (IR) fluorescence that replaces other near-IR dyes, including Cy5.5™ dye and Alexa Fluor™ 680, and is ideal for multi-color applications. The high water solubility of DyLight Fluors means that a high dye-to-protein ratio can be attained without causing precipitation of the conjugates. DyLight® 680 (Ex = 682 nm; Em = 715 nm)
This conjugate is suitable for immunomicroscopy and flow cytometry. The optimal working dilution should be determined by the investigator. Suggested starting dilution(?): · 1:20 - 1:2,000 for most applications
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 680
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hamster