The ImmunoReagents' DyLight 405 has a vibrant blue fluorescence intensity that replaces Alexa Fluor™ 405, Cascade™ Blue and AMCA in many applications. The high water solubility of DyLight Fluors means that a high dye-to-protein ratio can be attained without causing precipitation of the conjugates. DyLight® 405 (Ex = 400 nm; Em = 420 nm)
This antibody is suitable for all immunoassay applications. The optimal working dilution should be determined by the investigator.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat