Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Goat Anti-Mouse IgG Polyclonal Antibody (DyLight® 405)

Supplier: Immunoreagents
The ImmunoReagents' DyLight 405 has a vibrant blue fluorescence intensity that replaces Alexa Fluor™ 405, Cascade™ Blue and AMCA in many applications. The high water solubility of DyLight Fluors means that a high dye-to-protein ratio can be attained without causing precipitation of the conjugates. DyLight® 405 (Ex = 400 nm; Em = 420 nm)

This antibody is suitable for all immunoassay applications. The optimal working dilution should be determined by the investigator.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse



Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
1.0 mg IR2923 10013-980 Each (1mg) Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol IgG
Antigen Name Immunoglobulin G
Reactivity Mouse
Conjugation DyLight® 405
Clonality Polyclonal
Antibody Type Secondary
Host Goat
Isotype IgG
Western Blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Flow Cytometry Yes
Concentration 1.0 mg/ml (E 1% at 280 nm = 13.0)
Format Purified IgG
Cross Adsorption Yes
Immunogen Mouse IgG
Purification Affinity purified using solid phaseMouse IgG
Form Lyophilized
Storage Buffer 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free
Storage Temperature 2-8 °C
Shipping Temperature 2-8 °C
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty