|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | IgG |
Antigen Name | Immunoglobulin G |
Reactivity | Mouse |
Conjugation | HRP (Horseradish Peroxidase) |
Clonality | Polyclonal |
Antibody Type | Secondary |
Host | Goat |
Isotype | IgG |
Western Blot | Yes |
ELISA | Yes |
ImmunoChemistry | Yes |
ImmunoFluorescence | Yes |
Gel Shift | Yes |
Flow Cytometry | Yes |
Concentration | 1.0 mg/ml (E 1% at 280 nm = 13.0) |
Format | Purified IgG |
Cross Adsorption | Yes |
Immunogen | Mouse IgG |
Purification | Affinity purified using solid phaseMouse IgG |
Form | Lyophilized |
Storage Buffer | 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 1 % (w/v) BSA, Protease/IgG free |
Storage Temperature | 2-8 °C |
Shipping Temperature | 2-8 °C |