The ImmunoReagents' DyLight 405 has a vibrant blue fluorescence intensity that replaces Alexa Fluor™ 405, Cascade™ Blue and AMCA in many applications. The high water solubility of DyLight Fluors means that a high dye-to-protein ratio can be attained without causing precipitation of the conjugates. DyLight® 405 (Ex = 400 nm; Em = 420 nm)
This conjugate is suitable for immunomicroscopy and flow cytometry. The optimal working dilution should be determined by the investigator. Suggested starting dilution(?): · 1:20 - 1:2,000 for most applications
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 405
Epitope:
Host: Donkey
Isotype: IgG
Reactivity: Rat