Anti-CCL3 Rabbit Polyclonal Antibody

IR2764
10000-358EA 318.73 USD
10000-358
Anti-CCL3 Rabbit Polyclonal Antibody
Type: Primary
Antigen: CCL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR