Anti-IL27 Rabbit Polyclonal Antibody

IR2763
10000-356EA 318.73 USD
10000-356
Anti-IL27 Rabbit Polyclonal Antibody
Type: Primary
Antigen: IL27
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR