Anti-IL17F Rabbit Polyclonal Antibody

IR2761
10000-352EA 318.73 USD
10000-352
Anti-IL17F Rabbit Polyclonal Antibody
Type: Primary
Antigen: IL17F
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR