Anti-VEGF 121 Rabbit Polyclonal Antibody

IR2756
10000-342EA 318.73 USD
10000-342
Anti-VEGF 121 Rabbit Polyclonal Antibody
Type: Primary
Antigen: VEGF 121
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR