Anti-IL29 Rabbit Polyclonal Antibody

IR2750
10000-330EA 318.72 USD
10000-330
Anti-IL29 Rabbit Polyclonal Antibody
For antibody purification

Type: Primary
Antigen: IL29
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR