Anti-IL4 Rabbit Polyclonal Antibody

IR2743
10000-206EA 318.73 USD
10000-206
Anti-IL4 Rabbit Polyclonal Antibody
This antibody is suitable for all immunoassay applications. The optimal working dilution should be determined by the investigator.

Type: Primary
Antigen: IL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR