|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | GM-CSF |
Antigen Name | Granulocyte Macrophage Colony Stimulating Factor 1 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Rabbit |
Isotype | IgG |
Western Blot | Yes |
ELISA | Yes |
ImmunoChemistry | Yes |
ImmunoPrecipitation | Yes |
ImmunoFluorescence | Yes |
Flow Cytometry | Yes |
Antigen Synonyms | Molgramostin, CSF, Sargramostim, Granulocyte Macrophage Colony Stimulating Factor, Colony-stimulating factor |
Concentration | 0.5 mg/mL solution |
Format | Purified IgG |
Cross Adsorption | No |
Immunogen | Human GM-CSF |
Purification | Protein A |
Form | Lyophilized |
Storage Buffer | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with no stabilizers or preservatives |
Storage Temperature | 2-8 °C |
Shipping Temperature | 2-8 °C |