Anti-Interleukin-27 p28 Subunit Rat Monoclonal Antibody [clone: 3H12]

IR2722
10000-164EA 318.73 USD
10000-164
Anti-Interleukin-27 p28 Subunit Rat Monoclonal Antibody [clone: 3H12]
The p28 subunit of IL-27 (IL-27/p28), also known as Interleukin-30 (IL-30), is a member of the IL-12 family of cytokines.  When combined with EBI3 (Epstein-Barr virus induced gene 3), p28 forms a heterodimer known as IL-27.  p28 is thought to be a pro-inflammatory cytokine inducing immuno-modulatory effects, but the details of these effects are currently unclear and research is focused on delineating its specific biological functions. 

This conjugate is suitable for all immunoassay applications. The optimal working dilution should be determined by the investigator. Suggested starting dilution(?): · 1:500-1:5,000 for Immunohistochemistry · 1:200-1:5000 for ELISA/Western blot

Type: Primary
Antigen: Interleukin-27 p28 Subunit
Clonality: Monoclonal
Clone: 3H12
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR