2-Amino-6-chloro-4-nitrophenol 97%

Warning

Synonyms: 3-Chloro-2-hydroxy-5-nitroaniline

H66766-06 H66766-03 H66766-14
AAH66766-03EA 27.73 USD
AAH66766-03 AAH66766-14 AAH66766-06
2-Amino-6-chloro-4-nitrophenol 97%
2-Amino-6-chloro-4-nitrophenol
Formula: C₆H₅ClN₂O₃
MW: 188.57 g/mol
Melting Pt: 160…164 °C
Storage Temperature: Freezer
MDL Number: MFCD00035767
CAS Number: 6358-09-4
EINECS: 228-762-1
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR