2-Amino-4-chloro-6-nitrophenol 98%

Supplier: Thermo Scientific Chemicals

Warning

H66009-22 H66009-14
AAH66009-22EA 399.78 USD
AAH66009-22 AAH66009-14
2-Amino-4-chloro-6-nitrophenol 98%
2-Amino-4-chloro-6-nitrophenol
Formula: C₆H₅ClN₂O₃
Melting Pt: 158…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00035925
CAS Number: 6358-08-3
EINECS: 228-761-6
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR