N-Methylisopropylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Isopropylmethylamine

H65892-06 H65892-14
AAH65892-06EA 95.99 USD
AAH65892-06 AAH65892-14
N-Methylisopropylamine 98%
N-Methylisopropylamine
Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 50…53 °C
Density: 0.702
Flash Pt: -32 °C(-26 °F)
Storage Temperature: Ambient
MDL Number: MFCD00042859
CAS Number: 4747-21-1
EINECS: 225-266-7
UN: 2734
ADR: "8",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR