4-Methoxy-N-methylaniline 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
N-Methyl-p-anisidine
H65272-06
H65272-14
H65272-03
AAH65272-14EA
533.88
USD
AAH65272-14
AAH65272-03
AAH65272-06
4-Methoxy-N-methylaniline 96%
4-Methoxy-N-methylaniline
Formula:
C₈H₁₁NO MW: 137.18 g/mol Boiling Pt: 135…136 °C/19 mm Melting Pt: 33…36 °C Flash Pt: 110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008399 CAS Number: 5961-59-1 EINECS: 227-735-1 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...