N-Methoxy-N-methylacetamide 98%

Supplier: Thermo Scientific Chemicals
Warning

H65226-06 H65226-14 H65226-22
AAH65226-22EA 809.99 USD
AAH65226-22 AAH65226-06 AAH65226-14
N-Methoxy-N-methylacetamide 98%
N-Methoxy-N-methylacetamide
Formula: C4H9NO2
MW: 103.12 g/mol
Boiling Pt: 152 °C
Density: 0.970
Flash Pt: 49 °C(120 °F)
Storage Temperature: Ambient
MDL Number: MFCD00060098
CAS Number: 78191-00-1
UN: 1993
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR