5-Amino-2-fluoro-4-nitrobenzotrifluoride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Fluoro-2-nitro-5-(trifluoromethyl)aniline
H64928-03
H64928-06
H64928-MD
AAH64928-06EA
348.99
USD
AAH64928-06
AAH64928-MD
AAH64928-03
5-Amino-2-fluoro-4-nitrobenzotrifluoride 98%
5-Amino-2-fluoro-4-nitrobenzotrifluoride
Formula:
C₇H₄F₄N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD03094267 CAS Number: 179062-05-6 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...